Gene Transcripts

Gene Symbol
CDC7
Entrez Gene ID
8317
Gene Name
cell division cycle 7
Gene Aliases
CDC7L1, HsCDC7, Hsk1, huCDC7
Chromosome Location
Chr. 1: 91500814 - 91525764 on Build GRCh38
UniGene ID
Hs.533573
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134419.1 NP_001127891.1 2 215
NM_001134420.1 NP_001127892.1 2 343
NM_003503.3 NP_003494.1 2 249
XM_005271241.2 XP_005271298.1 2 211
XM_005271244.2 XP_005271301.1 2 249
XM_017002425.1 XP_016857914.1 2 211
XM_017002426.1 XP_016857915.1 2 194
GenBank mRNA AB003698.1 - 2 216
AB209337.1 - 1 925
AF005209.1 - 1 84
AF015592.1 - 2 229
AK298963.1 - 2 245
AK312726.1 - 2 305
BC110526.1 - 2 172 40034178
BC110527.1 - 2 206 40034179
BC111044.1 - 2 229 5518616
DA127303.1 - 2 343
DA791490.1 - 2 249

View Full Product Details