Gene Transcripts
- Gene Symbol
- C22orf39
- Entrez Gene ID
- 128977
- Gene Name
- chromosome 22 open reading frame 39
- Gene Aliases
- Chromosome Location
- Chr. 22: 19440886 - 19448232 on Build GRCh38
- UniGene ID
- Hs.659565
- Species
- Homo sapiens
- Gene Symbol
- HIRA
- Entrez Gene ID
- 7290
- Gene Name
- histone cell cycle regulator
- Gene Aliases
- DGCR1, TUP1, TUPLE1
- Chromosome Location
- Chr. 22: 19330701 - 19431696 on Build GRCh38
- UniGene ID
- Hs.474206
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK299950.1 |
- |
2 |
92 |
|
AK308642.1 |
- |
2 |
92 |
|
View Full Product Details