Gene Transcripts
- Gene Symbol
- KIR2DL4
- Entrez Gene ID
- 3805
- Gene Name
- killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 4
- Gene Aliases
- CD158D, G9P, KIR, KIR-103AS, KIR-2DL4, KIR103, KIR103AS
- Chromosome Location
- Chr. 19: 54803612 - 54814517 on Build GRCh38
- UniGene ID
- Hs.661219
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001080772.1 |
NP_001074241.1
|
5, 6 |
743 |
|
NM_002255.5 |
NP_002246.5
|
5, 6 |
743 |
|
GenBank mRNA |
AF002979.1 |
- |
5, 6 |
703 |
|
AF034771.1 |
- |
5, 6 |
709 |
|
AF034772.1 |
- |
5, 6 |
709 |
|
AF034773.1 |
- |
5, 6 |
709 |
|
AF276292.1 |
- |
5, 6 |
703 |
|
AF285436.1 |
- |
5, 6 |
730 |
|
AK309531.1 |
- |
6, 7 |
979 |
|
AY052496.1 |
- |
2, 3 |
274 |
|
AY052498.1 |
- |
2, 3 |
274 |
|
AY223512.2 |
- |
3, 4 |
630 |
|
AY223513.1 |
- |
3, 4 |
630 |
|
AY223514.1 |
- |
3, 4 |
630 |
|
AY250088.1 |
- |
3, 4 |
630 |
|
AY359817.1 |
- |
5, 6 |
714 |
|
AY601812.1 |
- |
5, 6 |
697 |
|
AY789058.1 |
- |
5, 6 |
697 |
|
AY789059.1 |
- |
5, 6 |
697 |
|
AY789060.1 |
- |
5, 6 |
697 |
|
AY789061.1 |
- |
5, 6 |
697 |
|
AY789062.1 |
- |
5, 6 |
697 |
|
BC041611.1 |
- |
5, 6 |
744 |
5485491 |
DQ266438.1 |
- |
5, 6 |
706 |
|
DQ272466.1 |
- |
3, 4 |
432 |
|
EU194340.1 |
- |
5, 6 |
703 |
|
EU194342.1 |
- |
5, 6 |
703 |
|
U71199.1 |
- |
5, 6 |
715 |
|
X97229.1 |
- |
5, 6 |
708 |
|
X99480.1 |
- |
5, 6 |
714 |
|
View Full Product Details