Gene Transcripts

Gene Symbol
KRTAP4-5
Entrez Gene ID
85289
Gene Name
keratin associated protein 4-5
Gene Aliases
KAP4.5, KRTAP4.5
Chromosome Location
Chr. 17: 41148924 - 41149802 on Build GRCh38
UniGene ID
Hs.514863
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033188.3 NP_149445.3 1 660
GenBank mRNA AJ406937.1 - 1 675

View Full Product Details