Gene Transcripts
- Gene Symbol
- TPPP2
- Entrez Gene ID
- 122664
- Gene Name
- tubulin polymerization promoting protein family member 2
- Gene Aliases
- C14orf8, CT152, P18, p25beta
- Chromosome Location
- Chr. 14: 21024262 - 21036352 on Build GRCh38
- UniGene ID
- Hs.406966
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_173846.4 |
NP_776245.2
|
2, 3 |
360 |
|
XM_005267324.4 |
XP_005267381.1
|
2, 3 |
421 |
|
XM_011536416.1 |
XP_011534718.1
|
2, 3 |
296 |
|
XM_011536417.2 |
XP_011534719.1
|
2, 3 |
296 |
|
XM_011536418.1 |
XP_011534720.1
|
2, 3 |
296 |
|
XM_011536420.2 |
XP_011534722.1
|
2, 3 |
296 |
|
XM_017020966.1 |
XP_016876455.1
|
2, 3 |
324 |
|
GenBank mRNA |
AY072034.1 |
- |
2, 3 |
297 |
|
AY173946.1 |
- |
2, 3 |
297 |
|
BC038970.2 |
- |
1, 2 |
249 |
5169309 |
View Full Product Details