Gene Transcripts

Gene Symbol
EFHC2
Entrez Gene ID
80258
Gene Name
EF-hand domain containing 2
Gene Aliases
MRX74, dJ1158H2.1
Chromosome Location
Chr. X: 44146254 - 44343677 on Build GRCh38
UniGene ID
Hs.521953
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025184.3 NP_079460.2 5 770
XM_006724562.2 XP_006724625.1 4 483
GenBank mRNA AK026496.1 - 5 623
AK291187.1 - 5 911
AK292967.1 - 5 754
AK297714.1 - 5 493
BC031039.1 - 5 765 5272398
CR749363.1 - 1 39

View Full Product Details