Gene Transcripts

Gene Symbol
LMAN2L
Entrez Gene ID
81562
Gene Name
lectin, mannose binding 2 like
Gene Aliases
MRT52, VIPL
Chromosome Location
Chr. 2: 96705929 - 96740092 on Build GRCh38
UniGene ID
Hs.655743
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142292.1 NP_001135764.1 1 191
NM_001322346.1 NP_001309275.1 1 207
NM_001322347.1 NP_001309276.1 1 207
NM_001322350.1 NP_001309279.1 1 207
NM_001322351.1 NP_001309280.1 1 207
NM_001322352.1 NP_001309281.1 1 207
NM_001322354.1 NP_001309283.1 1 207
NM_001322355.1 NP_001309284.1 1 207
NM_001322356.1 NP_001309285.1 1 207
NM_030805.3 NP_110432.1 1 191
GenBank mRNA AJ549957.1 - 1 155
AK075347.1 - 1 179
AK222828.1 - 1 169
AK295463.1 - 1 182
AK299740.1 - 1 179
AK301077.1 - 1 179
AK304515.1 - 1 178
AK308085.1 - 1 178
AK310100.1 - 1 208
AK312249.1 - 1 184
AK316331.1 - 1 179
AL136617.1 - 1 183
AY358929.1 - 1 162
BC000347.2 - 1 154
BC005822.2 - 1 154
BC005862.2 - 1 154
BC067265.1 - 1 161 6502212
BX648002.1 - 1 169
DA521101.1 - 1 168
DC354352.1 - 1 179

View Full Product Details