Gene Transcripts

Gene Symbol
ATCAY
Entrez Gene ID
85300
Gene Name
ATCAY, caytaxin
Gene Aliases
BNIP-H, CLAC
Chromosome Location
Chr. 19: 3880620 - 3928082 on Build GRCh38
UniGene ID
Hs.418055
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033064.4 NP_149053.1 4 604
GenBank mRNA AB058775.1 - 4 550
AK027889.1 - 4 539
AK092309.1 - 4 403
AK125457.1 - 4 247
AK299522.1 - 5 610
AY220297.1 - 3 174
BC026217.1 - 4 546 4153341
KU178814.1 - 3 174
KU178815.1 - 3 174

View Full Product Details