Gene Transcripts

Gene Symbol
VWA3A
Entrez Gene ID
146177
Gene Name
von Willebrand factor A domain containing 3A
Gene Aliases
Chromosome Location
Chr. 16: 22091930 - 22156966 on Build GRCh38
UniGene ID
Hs.10697
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173615.4 NP_775886.3 30 3265
XM_011545742.2 XP_011544044.1 34 3547
XM_011545744.2 XP_011544046.1 19 2147
XM_011545745.1 XP_011544047.1 17 1915
XM_011545746.2 XP_011544048.1 17 2018
XM_017022947.1 XP_016878436.1 35 3620
XM_017022948.1 XP_016878437.1 18 2719
XM_017022949.1 XP_016878438.1 16 2990
GenBank mRNA AK093467.1 - 6 828
AK128606.1 - 19 2133
AY360464.1 - 5 471
BC038400.1 - 7 710
BC109294.1 - 19 2040
BC109295.1 - 18 1993

View Full Product Details