Gene Transcripts
- Gene Symbol
- NUSAP1
- Entrez Gene ID
- 51203
- Gene Name
- nucleolar and spindle associated protein 1
- Gene Aliases
- ANKT, BM037, LNP, NUSAP, PRO0310p1, Q0310, SAPL
- Chromosome Location
- Chr. 15: 41332694 - 41381050 on Build GRCh38
- UniGene ID
- Hs.615092
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001243142.1 |
NP_001230071.1
|
1, 2 |
364 |
|
NM_001243143.1 |
NP_001230072.1
|
1, 2 |
364 |
|
NM_001301136.1 |
NP_001288065.1
|
1, 2 |
364 |
|
NM_016359.4 |
NP_057443.2
|
1, 2 |
364 |
|
NM_018454.7 |
NP_060924.4
|
1, 2 |
364 |
|
XM_005254428.2 |
XP_005254485.1
|
1, 2 |
218 |
|
XM_005254430.4 |
XP_005254487.1
|
1, 2 |
203 |
|
XM_005254431.2 |
XP_005254488.1
|
1, 2 |
218 |
|
XM_006720559.2 |
XP_006720622.1
|
1, 2 |
218 |
|
XM_006720560.3 |
XP_006720623.1
|
1, 2 |
317 |
|
XM_006720561.2 |
XP_006720624.1
|
1, 2 |
177 |
|
XM_006720562.2 |
XP_006720625.1
|
1, 2 |
218 |
|
XM_006720563.2 |
XP_006720626.1
|
1, 2 |
218 |
|
XM_017022294.1 |
XP_016877783.1
|
1, 2 |
203 |
|
XM_017022295.1 |
XP_016877784.1
|
1, 2 |
153 |
|
GenBank mRNA |
AF090915.1 |
- |
1, 2 |
168 |
|
AF217513.1 |
- |
1, 2 |
178 |
|
AF290612.1 |
- |
1, 2 |
168 |
|
AF305711.1 |
- |
1, 2 |
100 |
|
AK023483.1 |
- |
1, 2 |
174 |
|
AK222819.1 |
- |
1, 2 |
188 |
|
AK225703.1 |
- |
1, 2 |
175 |
|
BC001308.1 |
- |
1, 2 |
167 |
3453354 |
BC012887.1 |
- |
1, 2 |
145 |
3882061 |
BC024772.1 |
- |
1, 2 |
172 |
4342499 |
View Full Product Details