Gene Transcripts

Gene Symbol
SULT1A2
Entrez Gene ID
6799
Gene Name
sulfotransferase family 1A member 2
Gene Aliases
HAST4, P-PST, ST1A2, STP2, TSPST2
Chromosome Location
Chr. 16: 28591943 - 28597079 on Build GRCh38
UniGene ID
Hs.546304
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001054.3 NP_001045.1 8 1029
NM_177528.2 NP_803564.1 8 1302
XM_006721077.3 XP_006721140.1 4 1566
XM_017023600.1 XP_016879089.1 8 1056
XM_017023601.1 XP_016879090.1 8 1484
XM_017023602.1 XP_016879091.1 9 1186
XM_017023603.1 XP_016879092.1 6 812
GenBank mRNA U28169.1 - 8 1024
U28170.1 - 8 1012

View Full Product Details