Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000207.2 | NP_000198.1 | 2 | 179 | |
NM_001185097.1 | NP_001172026.1 | 2 | 205 | ||
NM_001185098.1 | NP_001172027.1 | 1 | 358 | ||
NM_001291897.1 | NP_001278826.1 | 2 | 239 | ||
GenBank mRNA | AY899304.1 | - | 2 | 175 | |
BC005255.1 | - | 2 | 179 | 3950204 | |
BM510347.1 | - | 2 | 200 | ||
BM510748.1 | - | 2 | 183 | ||
BP322143.1 | - | 1 | 358 | ||
BT006808.1 | - | 1 | 120 | ||
DQ778082.1 | - | 1 | 120 | ||
JF909299.1 | - | 1 | 73 | ||
JQ951950.1 | - | 2 | 164 | ||
X70508.1 | - | 2 | 164 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001042376.2 | NP_001035835.1 | 2 | 179 | |
NR_003512.3 | 2 | 179 | |||
GenBank mRNA | DQ104204.1 | - | 2 | 179 | |
DQ104205.1 | - | 2 | 179 |