Gene Transcripts

Gene Symbol
UBE2D2
Entrez Gene ID
7322
Gene Name
ubiquitin conjugating enzyme E2 D2
Gene Aliases
E2(17)KB2, PUBC1, UBC4, UBC4/5, UBCH4, UBCH5B
Chromosome Location
Chr. 5: 139561166 - 139628434 on Build GRCh38
UniGene ID
Hs.108332
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003339.2 NP_003330.1 2, 3 716
NM_181838.1 NP_862821.1 3, 4 880
XM_017009820.1 XP_016865309.1 5, 6 1087
GenBank mRNA AF317220.1 - 24
AK001311.1 - 2, 3 489
AK001428.1 - 2, 3 543
AK310273.1 - 2, 3 445
AY651263.1 - 2, 3 472
BC019052.2 - 2, 3 500
BC033349.1 - 2, 3 716 4827210
BQ186099.1 - 3, 4 365
U39317.1 - 2, 3 112

View Full Product Details