Gene Transcripts

Gene Symbol
NIT2
Entrez Gene ID
56954
Gene Name
nitrilase family member 2
Gene Aliases
HEL-S-8a
Chromosome Location
Chr. 3: 100334718 - 100355634 on Build GRCh38
UniGene ID
Hs.439152
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020202.4 NP_064587.1 2 188
GenBank mRNA AF260334.1 - 2 187
AF284574.1 - 2 135
AK225979.1 - 2 164
AK294010.1 - 2 149
AK295835.1 - 2 149
AK313704.1 - 2 130
BC020620.1 - 2 145 4775062
BC107890.1 - 2 176 6648910
DB466042.1 - 2 188
FJ224326.1 - 2 135

View Full Product Details