Gene Transcripts

Gene Symbol
Rap2a
Entrez Gene ID
114560
Gene Name
RAS related protein 2a
Gene Aliases
Chromosome Location
Chr. 15: 105851543 - 105852033 on Build Rnor_6.0
UniGene ID
Rn.11606
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053741.1 NP_446193.1 1 441
GenBank mRNA AY037826.1 - 1 441

View Full Product Details