Gene Transcripts

Gene Symbol
Banf1
Entrez Gene ID
114087
Gene Name
barrier to autointegration factor 1
Gene Aliases
Baf, Bcrp1, L2bp1, L2bp1/Baf
Chromosome Location
Chr. 1: 220744195 - 220746224 on Build Rnor_6.0
UniGene ID
Rn.19921
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053631.3 NP_446083.1 3 264
XM_006230630.2 XP_006230692.1 3 277
XM_006230631.1 XP_006230693.1 3 301
XM_006230632.2 XP_006230694.1 3 274
GenBank mRNA AB024333.1 - 3 259
BC084726.1 - 3 235 7320690
CB766065.1 - 3 264
FQ224698.1 - 3 306

View Full Product Details