Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012796.2 | NP_036928.1 | 4 | 535 | |
GenBank mRNA | BC061856.1 | - | 4 | 539 | |
CK473092.1 | - | 4 | 535 | ||
D10026.1 | - | 4 | 523 | ||
FJ179409.1 | - | 4 | 401 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_008773848.1 | XP_008772070.1 | 2 | 605 |