Gene Transcripts

Gene Symbol
Hal
Entrez Gene ID
29301
Gene Name
histidine ammonia lyase
Gene Aliases
histidase
Chromosome Location
Chr. 7: 34326087 - 34356413 on Build Rnor_6.0
UniGene ID
Rn.10037
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017159.1 NP_058855.1 1 241
GenBank mRNA BC089809.1 - 1 270
M58308.1 - 1 241

View Full Product Details