Gene Transcripts

Gene Symbol
Klhl3
Entrez Gene ID
100503085
Gene Name
kelch-like 3
Gene Aliases
7530408C15Rik, AI430941, EG627648
Chromosome Location
Chr. 13: 58004439 - 58123481 on Build GRCm38
UniGene ID
Mm.379256
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195075.1 NP_001182004.1 5 319

View Full Product Details