Gene Transcripts

Gene Symbol
Rprml
Entrez Gene ID
104582
Gene Name
reprimo-like
Gene Aliases
AW049332
Chromosome Location
Chr. 11: 103649509 - 103650580 on Build GRCm38
UniGene ID
Mm.44618
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033212.2 NP_001028384.1 1 612
GenBank mRNA AK141598.1 - 1 613
BC147064.1 - 1 587 9056034
BC147065.1 - 1 587 9056062

View Full Product Details