Gene Transcripts

Gene Symbol
Itga1
Entrez Gene ID
109700
Gene Name
integrin alpha 1
Gene Aliases
CD49A, E130012M19Rik, Vla1
Chromosome Location
Chr. 13: 114958079 - 115101964 on Build GRCm38
UniGene ID
Mm.317280
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033228.3 NP_001028400.2 26, 27 3713
GenBank mRNA AK035870.1 - 26, 27 3714
AK040830.1 - 17, 18 2137
AK053377.1 - 1, 2 112

View Full Product Details