Gene Transcripts

Gene Symbol
Crygf
Entrez Gene ID
12969
Gene Name
crystallin, gamma F
Gene Aliases
3110001K11Rik, Cryg-2, DGcry-2, Len-2
Chromosome Location
Chr. 1: 65926522 - 65928304 on Build GRCm38
UniGene ID
Mm.275362
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027010.2 NP_081286.1 3 590
GenBank mRNA AK013953.1 - 3 591
BC056453.1 - 3 581 6824171

View Full Product Details