Gene Transcripts

Gene Symbol
Kif7
Entrez Gene ID
16576
Gene Name
kinesin family member 7
Gene Aliases
Chromosome Location
Chr. 7: 79698098 - 79715772 on Build GRCm38
UniGene ID
Mm.101633
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291222.1 NP_001278151.1 10 2396
NM_010626.3 NP_034756.2 10 2396
GenBank mRNA BC141045.1 - 10 2397
BC145222.1 - 10 2397 9053773

View Full Product Details