Gene Transcripts

Gene Symbol
Lrrc73
Entrez Gene ID
224813
Gene Name
leucine rich repeat containing 73
Gene Aliases
Gm88
Chromosome Location
Chr. 17: 46253445 - 46257316 on Build GRCm38
UniGene ID
Mm.70999
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001111142.1 NP_001104612.1 6 1359
GenBank mRNA AK134769.1 - 4 2425

View Full Product Details