Gene Transcripts

Gene Symbol
Gm94
Entrez Gene ID
225443
Gene Name
predicted gene 94
Gene Aliases
Chromosome Location
Chr. 18: 43777196 - 43792878 on Build GRCm38
UniGene ID
Mm.100304
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033280.2 NP_001028452.1 2 215
GenBank mRNA AK131907.1 - 2 216
AK131908.1 - 2 216
BC147098.1 - 2 204 9056146
BC147099.1 - 2 204 9056134
BC171995.1 - 2 204 9054281

View Full Product Details