Gene Transcripts

Gene Symbol
Slc30a10
Entrez Gene ID
226781
Gene Name
solute carrier family 30, member 10
Gene Aliases
E130106K10Rik, Gm212
Chromosome Location
Chr. 1: 185454848 - 185468762 on Build GRCm38
UniGene ID
Mm.227117
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033286.2 NP_001028458.1 4 5418
GenBank mRNA AK046969.1 - 1 1434
BC080293.1 - 2 4493
BC108419.1 - 4 4804

View Full Product Details