Gene Transcripts

Gene Symbol
Chd5
Entrez Gene ID
269610
Gene Name
chromodomain helicase DNA binding protein 5
Gene Aliases
4930532L22Rik, AW060752, B230399N07Rik, CHD-5
Chromosome Location
Chr. 4: 152338651 - 152390194 on Build GRCm38
UniGene ID
Mm.40192
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081376.1 NP_001074845.1 14 2434
NM_029216.2 NP_083492.2 14 2434
GenBank mRNA AK147563.1 - 14 2435

View Full Product Details