Gene Transcripts

Gene Symbol
Tmem88b
Entrez Gene ID
320587
Gene Name
transmembrane protein 88B
Gene Aliases
A230069A22Rik
Chromosome Location
Chr. 4: 155781591 - 155785874 on Build GRCm38
UniGene ID
Mm.384382
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033394.3 NP_001028566.2 2 1246
GenBank mRNA AK035998.1 - 2 1246
AK038855.1 - 151
AK158459.1 - 2 1247

View Full Product Details