Gene Transcripts

Gene Symbol
Sftpc
Entrez Gene ID
50683
Gene Name
surfactant protein C
Gene Aliases
SP-C
Chromosome Location
Chr. 15: 52211538 - 52214480 on Build Rnor_6.0
UniGene ID
Rn.3926
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017342.2 NP_059038.1 4 374
GenBank mRNA BC072693.1 - 4 367
X14221.1 - 4 375

View Full Product Details