Gene Transcripts

Gene Symbol
Ppp1r12b
Entrez Gene ID
329251
Gene Name
protein phosphatase 1, regulatory (inhibitor) subunit 12B
Gene Aliases
1810037O03Rik, 9530009M10Rik, AI132431, Mypt2
Chromosome Location
Chr. 1: 134754665 - 134955965 on Build GRCm38
UniGene ID
Mm.188709
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081307.1 NP_001074776.1 16 2419
GenBank mRNA AK039445.1 - 4 480
AK141375.1 - 18 2634
BC145746.1 - 16 2330 40131049

View Full Product Details