Gene Transcripts

Gene Symbol
Birc7
Entrez Gene ID
329581
Gene Name
baculoviral IAP repeat-containing 7 (livin)
Gene Aliases
E130019N06, KIAP, Livin, ML-IAP
Chromosome Location
Chr. 2: 180928152 - 180935028 on Build GRCm38
UniGene ID
Mm.187879
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163247.1 NP_001156719.1 5 773
GenBank mRNA AK053463.1 - 4 1127
BC107260.2 - 5 697
BC125013.1 - 5 697

View Full Product Details