Gene Transcripts

Gene Symbol
Kif14
Entrez Gene ID
381293
Gene Name
kinesin family member 14
Gene Aliases
D1Ertd367e, E130203M01
Chromosome Location
Chr. 1: 136466290 - 136555938 on Build GRCm38
UniGene ID
Mm.458468
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287179.2 NP_001274108.1 2 1408
GenBank mRNA AB731728.1 - 2 1298
AK087451.1 - 3 1437
BC158132.1 - 2 1321 9007399

View Full Product Details