Gene Transcripts

Gene Symbol
Cracr2a
Entrez Gene ID
381812
Gene Name
calcium release activated channel regulator 2A
Gene Aliases
Efcab4b, Gm1073, Gm462
Chromosome Location
Chr. 6: 127561064 - 127670161 on Build GRCm38
UniGene ID
Mm.296093
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033464.3 NP_001028636.1 5 789
GenBank mRNA AK143832.1 - 5 790
BC150869.1 - 5 867 9087780

View Full Product Details