Gene Transcripts
- Gene Symbol
- Spt1
- Entrez Gene ID
- 29229
- Gene Name
- salivary protein 1
- Gene Aliases
- Chromosome Location
- Chr. 7: 145325004 - 145338152 on Build Rnor_6.0
- UniGene ID
- Rn.9933
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019171.1 |
NP_062044.1
|
3, 4 |
269 |
|
GenBank mRNA |
M33976.1 |
- |
3, 4 |
269 |
|
View Full Product Details