Gene Transcripts

Gene Symbol
Tmem29
Entrez Gene ID
382245
Gene Name
transmembrane protein 29
Gene Aliases
2700081K05Rik, 6330540D07Rik, CXorf44-like
Chromosome Location
Chr. X: 150397773 - 150467331 on Build GRCm38
UniGene ID
Mm.277309
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164683.1 NP_001158155.1 3 1241
NM_001164684.1 NP_001158156.1 3 1210
GenBank mRNA AK018228.1 - 3 1206

View Full Product Details