Gene Transcripts

Gene Symbol
Fam47e
Entrez Gene ID
384198
Gene Name
family with sequence similarity 47, member E
Gene Aliases
C730043O17, Gm1381
Chromosome Location
Chr. 5: 92554951 - 92591285 on Build GRCm38
UniGene ID
Mm.76757
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033478.2 NP_001028650.2 3, 4 430
GenBank mRNA AK144479.1 - 3, 4 430

View Full Product Details