Gene Transcripts

Gene Symbol
Zyg11b
Entrez Gene ID
414872
Gene Name
zyg-ll family member B, cell cycle regulator
Gene Aliases
1110046I03Rik, 2810482G21Rik, D4Mgi23, Gm431, mKIAA1730
Chromosome Location
Chr. 4: 108227755 - 108301141 on Build GRCm38
UniGene ID
Mm.426241
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033634.3 NP_001028806.2 14 7949
GenBank mRNA AK004185.1 - 1 156
AK034007.1 - 1 356
AK036367.1 - 1 109
AK122545.1 - 7 6338

View Full Product Details