Gene Transcripts

Gene Symbol
Khdc1c
Entrez Gene ID
433278
Gene Name
KH domain containing 1C
Gene Aliases
Khdc1b
Chromosome Location
Chr. 1: 21368331 - 21369743 on Build GRCm38
UniGene ID
Mm.26006
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033904.1 NP_001029076.1 1 27
GenBank mRNA BC099381.1 - 1 27

View Full Product Details