Gene Transcripts

Gene Symbol
Vmn2r9
Entrez Gene ID
435864
Gene Name
vomeronasal 2, receptor 9
Gene Aliases
EG435864
Chromosome Location
Chr. 5: 108842947 - 108852510 on Build GRCm38
UniGene ID
Mm.387745
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104621.1 NP_001098091.1 3 1255

View Full Product Details