Gene Transcripts

Gene Symbol
Scrt2
Entrez Gene ID
545474
Gene Name
scratch homolog 2, zinc finger protein (Drosophila)
Gene Aliases
AW060659
Chromosome Location
Chr. 2: 152081529 - 152095802 on Build GRCm38
UniGene ID
Mm.214317
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160410.1 NP_001153882.1 2 2740
GenBank mRNA AK090280.1 - 1, 2 1794

View Full Product Details