Gene Transcripts

Gene Symbol
Vmn2r73
Entrez Gene ID
620928
Gene Name
vomeronasal 2, receptor 73
Gene Aliases
EG620928
Chromosome Location
Chr. 7: 85857547 - 85875938 on Build GRCm38
UniGene ID
Mm.469906
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105186.1 NP_001098656.1 3 586

View Full Product Details