Gene Transcripts

Gene Symbol
Vmn2r79
Entrez Gene ID
621430
Gene Name
vomeronasal 2, receptor 79
Gene Aliases
EG621430
Chromosome Location
Chr. 7: 86996465 - 87037968 on Build GRCm38
UniGene ID
Mm.469903
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105190.1 NP_001098660.1 6 1684

View Full Product Details