Gene Transcripts

Gene Symbol
Lekr1
Entrez Gene ID
624866
Gene Name
leucine, glutamate and lysine rich 1
Gene Aliases
EG546798, EG624866, Gm6534
Chromosome Location
Chr. 3: 65659539 - 65832480 on Build GRCm38
UniGene ID
Mm.296317
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166659.1 NP_001160131.1 9 1048

View Full Product Details