Gene Transcripts

Gene Symbol
Gcnt7
Entrez Gene ID
654821
Gene Name
glucosaminyl (N-acetyl) transferase family member 7
Gene Aliases
A330041C17Rik, gnt
Chromosome Location
Chr. 2: 172447120 - 172458596 on Build GRCm38
UniGene ID
Mm.389337
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039560.3 NP_001034649.1 2 858
GenBank mRNA AK039414.1 - 2 860
AK132647.1 - 1 531
BC139073.1 - 2 818 8862095
BC139076.1 - 2 818 8862098

View Full Product Details