Gene Transcripts

Gene Symbol
Cenpw
Entrez Gene ID
66311
Gene Name
centromere protein W
Gene Aliases
2610036L11Rik, Cug2
Chromosome Location
Chr. 10: 30196009 - 30200600 on Build GRCm38
UniGene ID
Mm.251779
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109747.1 NP_001103217.1 3 1135
GenBank mRNA AK011700.1 - 3 1116
AK141272.1 - 3 1117
BC050071.1 - 2326

View Full Product Details