Gene Transcripts

Gene Symbol
Cwc27
Entrez Gene ID
67285
Gene Name
CWC27 spliceosome-associated protein homolog (S. cerevisiae)
Gene Aliases
3110009E13Rik, NY-CO-10, Sdccag10
Chromosome Location
Chr. 13: 104631322 - 104816964 on Build GRCm38
UniGene ID
Mm.235398
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026072.1 NP_080348.1 3 389
GenBank mRNA AK014025.1 - 3 373
AK075902.1 - 3 367
AK077779.1 - 3 332
AK166770.1 - 3 389
BC025437.1 - 3 189 5025084

View Full Product Details