Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026364.1 | NP_080640.1 | 10 | 1179 | |
GenBank mRNA | AK011290.1 | - | 10 | 1149 | |
AK077439.1 | - | 10 | 1182 | ||
AK082484.1 | - | 9 | 1033 | ||
AK178677.1 | - | 2 | 113 | ||
AK181341.1 | - | 2 | 181 | ||
AK182019.1 | - | 2 | 181 | ||
AK200075.1 | - | 2 | 181 | ||
BC029621.1 | - | 10 | 1170 | 4208743 |