Gene Transcripts

Gene Symbol
Kctd17
Entrez Gene ID
72844
Gene Name
potassium channel tetramerisation domain containing 17
Gene Aliases
2900008M13Rik, AA414907, AW742389, N28155
Chromosome Location
Chr. 15: 78428559 - 78439303 on Build GRCm38
UniGene ID
Mm.390816
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289671.1 NP_001276600.1 8 1248
NM_001289672.1 NP_001276601.1 7 1173
NM_001289673.1 NP_001276602.1 6 1085
GenBank mRNA AK013500.1 - 1 100

View Full Product Details