Gene Transcripts

Gene Symbol
Ankrd45
Entrez Gene ID
73844
Gene Name
ankyrin repeat domain 45
Gene Aliases
4933409K03Rik
Chromosome Location
Chr. 1: 161142547 - 161170507 on Build GRCm38
UniGene ID
Mm.38830
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028664.1 NP_082940.1 3 602
GenBank mRNA AK016757.1 - 2 521
BC049713.1 - 3 574 6741553

View Full Product Details