Gene Transcripts

Gene Symbol
Arhgap36
Entrez Gene ID
75404
Gene Name
Rho GTPase activating protein 36
Gene Aliases
1100001E04Rik, rho-gap
Chromosome Location
Chr. X: 49448328 - 49500250 on Build GRCm38
UniGene ID
Mm.44305
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081123.1 NP_001074592.1 13 2651
GenBank mRNA AK048167.1 - 13 2540
BC090672.1 - 13 2622 30941825

View Full Product Details